Tantalum(V) oxide ≥99.95%

Supplier: BeanTown Chemical

Synonyms: Ditantalum pentaoxide

224745-100G 224745-25G 224745-500G
BT224745-100GEA 438 USD
BT224745-100G BT224745-25G BT224745-500G
Tantalum(V) oxide ≥99.95%
Tantalum (V) oxide

CAS: 1314-61-0; EC No: 215-238-2; MDL No: MFCD00011254; RTECS: WW5855000 Powder; Linear Formula: Ta2O5; MW: 441.89 Melting Point: 1800° Density (g/mL): 8.2


  • RTECS: WW5855000
  • Powder

Formula: O₅Ta₂
Melting Pt: 1800 °C
Density: 8.2 g/mL
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR