Ytterbium ≥99.9% (trace rare earth metals basis), ingot

Supplier: BeanTown Chemical

224710-25G 224710-5G
BT224710-25GEA 343 USD
BT224710-25G BT224710-5G
Ytterbium ≥99.9% (trace rare earth metals basis), ingot
Ytterbium

CAS: 7440-64-4; EC No: 231-173-2; MDL No: MFCD00011286 Solid, packaged in air stable poly bottle; Molecular Formula: Yb; MW: 173.04 Melting Point: 824°; Boiling Point: 1196° Density (g/mL): 6.54


  • Solid, packaged in air stable poly bottle

Formula: Yb
MW: 173.04 g/mol
Boiling Pt: 1427
Melting Pt: 824
Density: 6.54 g/mL
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR