L(+)-Tartaric acid ACS
Supplier: BeanTown Chemical
Synonyms:
L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
224505-100G
224505-2KG
224505-500G
BT224505-100GEA
37.96
USD
BT224505-100G
BT224505-2KG
BT224505-500G
L(+)-Tartaric acid ACS
L(+)-Tartaric acid
CAS: 87-69-4; EC No: 201-766-0; MDL No: MFCD00064207; RTECS: WW7875000
Granular; Molecular Formula: C4H6O6; MW: 150.09
Melting Point: 170-172°
Density (g/mL): 1.76
- RTECS: WW7875000
- Granular
Formula:
C₄H₆O₆ MW: 150.09 g/mol Boiling Pt: 399 Melting Pt: 170…172 °C Density: 1.76 g/mL Flash Pt: 150 |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 EINECS: 201-766-0 Merck Index: 13,09158 |
|
Specification Test Results
Loss on drying | ≤0.05% |
Residue after ignition | ≤0.005% |
Oxalate (C2O4) | ≤0.1% |
Sulfur compounds | ≤0.002% |
Heavy metals (as Pb) | ≤5ppm |
Chloride (Cl) | ≤0.001% |
Insoluble matter | ≤0.005% |
Phosphate (PO4) | ≤0.001% |
Iron (Fe) | ≤5ppm |
Specific rotation | +12.40 deg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...