Citric acid monohydrate 99.0-102.0% ACS
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
224230-100G
224230-2KG
224230-500G
BT224230-100GEA
30.01
USD
BT224230-100G
BT224230-2KG
BT224230-500G
Citric acid monohydrate 99.0-102.0% ACS
Citric acid monohydrate
CAS: 5949-29-1; EC No: 201-069-1; MDL No: MFCD00149972; RTECS: GE7810000
Solid; Molecular Formula: C6H8O7·H2O ; MW: 210.14
Density (g/mL): 1.542
Hygroscopic
- RTECS: GE7810000
- Hygroscopic
- Solid
Formula:
C₆H₈O₇.H₂O MW: 210.14 g/mol Density: 1.542 g/mL Flash Pt: 173.9 |
MDL Number:
MFCD00149972 CAS Number: 5949-29-1 EINECS: 201-069-1 Merck Index: 13,02350 |
|
Specification Test Results
Assay | 99.0-102.0% |
Insoluble matter | ≤0.005% |
Residue after ignition | ≤0.02% |
Chloride (Cl) | ≤0.001% |
Oxalate (C2O4) | Passes test |
Phosphate (PO4) | ≤0.001% |
Sulfate (SO4) | ≤0.002% |
Iron (Fe) | ≤3ppm |
Lead (Pb) | ≤2ppm |
Substances carbonizable by hot sulfuric acid | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...