(R)-(-)-3-Aminopiperidine dihydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
(R)-(-)-3-Piperidinamine dihydrochloride, (R)-Piperidin-3-amine dihydrochloride
223995-1G
223995-250MG
BT223995-1GEA
123
USD
BT223995-1G
BT223995-250MG
(R)-(-)-3-Aminopiperidine dihydrochloride ≥98%
(R)-(-)-3-Aminopiperidine dihydrochloride
CAS: 334618-23-4; MDL No: MFCD06799458
Powder; Molecular Formula: C5H14Cl2N2; MW: 173.08
Melting Point: 190-195°
Optical Rotation: [α]/D -2.0±0.5°, c = 1 in water
Hygroscopic
- Optical rotation: [α]/D -2.0±0.5°, c = 1 in water
- Hygroscopic
- Powder
Formula:
C₅H₁₄Cl₂N₂ Melting Pt: 190…195 °C |
MDL Number:
MFCD06799458 CAS Number: 334618-23-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...