Zinc molybdate ≥98%

Supplier: BeanTown Chemical

Synonyms: Zinc molybdenum oxide

223930-25G 223930-5G
BT223930-25GEA 246 USD
BT223930-25G BT223930-5G
Zinc molybdate ≥98%
Zinc molybdate

CAS: 13767-32-3; EC No: 237-377-8; MDL No: MFCD00049622 Powder; Linear Formula: ZnMoO4; MW: 225.32 Melting Point: 700° Density (g/mL): 4.300


  • Powder

Formula: MoO₄Zn
MW: 225.326 g/mol
Melting Pt: >700 °C
Density: 4.300 g/mL
MDL Number: MFCD00049622
CAS Number: 13767-32-3
EINECS: 237-377-8

Order Now

Specification Test Results

Assay 98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR