p-Toluenesulfinic Acid Sodium Salt ≥97% (dry weight)
Supplier: BeanTown Chemical
223920-100G
223920-25G
223920-500G
BT223920-100GEA
47.78
USD
BT223920-100G
BT223920-25G
BT223920-500G
p-Toluenesulfinic Acid Sodium Salt ≥97% (dry weight)
p-Toluenesulfinic Acid Sodium Salt
CAS: 824-79-3; EC No: 212-538-5; MDL No: MFCD00149640; RTECS: XT4725000
Powder; Linear Formula: CH3C6H4SO2Na; Molecular Formula: C7H7NaO2S; MW: 178.19
Melting Point:
- RTECS: XT4725000
- Hygroscopic
Formula:
C₇H₇NaO₂S Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013136 CAS Number: 824-79-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...