5-Methylphenazinium methyl sulfate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Phenazine methosulfate, PMS, 5-Methylphenazin-5-ium methyl sulfate, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulfate, Phenazine methylsulphate
223820-25G
223820-5G
BT223820-25GEA
393
USD
BT223820-25G
BT223820-5G
5-Methylphenazinium methyl sulfate ≥98%
5-Methylphenazinium methyl sulphate
CAS: 299-11-6; EC No: 206-072-1; MDL No: MFCD00011923; RTECS: SG1645000
Crystalline/Powder; Linear Formula: C13H11N2·CH3OSO3; Molecular Formula: C13H11N2·CH3O4S; MW: 306.34
Melting Point: 158-160° (decomposes)
Light Sensitive
- RTECS: SG1645000
- Light Sensitive
- Crystalline/Powder
Formula:
C₁₄H₁₄N₂O₄S MW: 306.34 g/mol Melting Pt: 198 (dec.) |
MDL Number:
MFCD00011923 CAS Number: 299-11-6 EINECS: 206-072-1 Merck Index: 12,06185 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...