Gold(III) acetate ≥99.9%
Supplier: BeanTown Chemical
223795-1G
223795-500MG
223795-5G
BT223795-1GEA
219
USD
BT223795-1G
BT223795-500MG
BT223795-5G
Gold(III) acetate ≥99.9%
Gold (III) acetate
CAS: 15804-32-7; MDL No: MFCD00210614
Powder; Linear Formula: Au(O2CCH3)3; Molecular Formula: C6H9AuO6; MW: 374.10
- Powder
Formula:
C₆H₉AuO₆ MW: 374.10 g/mol |
MDL Number:
MFCD00210614 CAS Number: 15804-32-7 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...