Polyethylene glycol monomethyl ether, M.W. ∼350

Supplier: BeanTown Chemical

Synonyms: Methoxypolyethylene glycols-average M.W. 1000mPEGMethoxy poly(ethylene glycol)Methoxypolyethylene glycolsm-PEG-OHPEG MMEPoly(ethylene glycol) methyl ether

223750-100G 223750-500G
BT223750-500GEA 128.39 USD
BT223750-500G BT223750-100G
Polyethylene glycol monomethyl ether, M.W. ∼350
Polyethylene glycol monomethyl ether

  • Liquid

Formula: CH₃(OCH₂CH₂)nOH
Melting Pt: –8 °C
Density: 1.094 g/mL
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR