Folic acid ≥96%

Supplier: BeanTown Chemical

Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

223575-100G 223575-25G
BT223575-100GEA 204 USD
BT223575-100G BT223575-25G
Folic acid ≥96%
Folic acid

CAS: 59-30-3; EC No: 200-419-0; MDL No: MFCD00014787; RTECS: LP5425000 Powder; Molecular Formula: C15H19N7O6; MW: 441.40


  • RTECS: LP5425000
  • Powder

Formula: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR