(-)-Quinine, anhydrous ≥99%
Supplier: BeanTown Chemical
CAS: 130-95-0; EC No: 205-003-2; MDL No: MFCD00198096; RTECS: VA6020000
Crystalline; Molecular Formula: C20H24N2O2; MW: 324.42
Melting Point: 173-175°
Optical Rotation: [α]20/D -126±5°, c = 1% in chloroform
Light Sensitive
- RTECS: VA6020000
- Optical rotation: [α]20/D -126±5°, c = 1% in chloroform
- Light Sensitive
- Crystalline
Formula:
C₂₀H₂₄N₂O₂ Melting Pt: 173…175 °C |
MDL Number:
MFCD00198096 CAS Number: 130-95-0 EINECS: 205-003-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...