4-Amino-3-nitropyridine ≥96%

Supplier: BeanTown Chemical

Synonyms: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

223210-1G 223210-25G 223210-5G
BT223210-1GEA 32 USD
BT223210-1G BT223210-25G BT223210-5G
4-Amino-3-nitropyridine ≥96%
4-Amino-3-nitropyridine

CAS: 1681-37-4; MDL No: MFCD00160304 Crystalline/Powder; Molecular Formula: C5H5N3O2 ; MW: 139.11 Melting Point: 203-207°


  • Crystalline/Powder

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 203…207 °C
MDL Number: MFCD00160304
CAS Number: 1681-37-4

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR