Tin ≥98.5% (trace metals basis), foil, Thickness 0.05 mm
Supplier: BeanTown Chemical
223085-15X15CM
223085-15X300
223085-15X75CM
BT223085-15X15EA
73
USD
BT223085-15X15
BT223085-15X30
BT223085-15X75
Tin ≥98.5% (trace metals basis), foil, Thickness 0.05 mm
Tin
CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862
Solid; Molecular Formula: Sn; MW: 118.71
Melting Point: 231.9°; Boiling Point: 2270°
Density (g/mL): 7.31
- Solid
Formula:
Sn Boiling Pt: 2270 °C Melting Pt: 231.9 °C Density: 7.31 g/mL |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 |
Specification Test Results
Assay | 98.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...