Platinum ≥99.99% (trace metals basis), foil, Thickness 0.05 mm

Supplier: BeanTown Chemical

223030-25X25MM 223030-50X50MM
BT223030-25X25EA 276.37 USD
BT223030-25X25 BT223030-50X50
Platinum ≥99.99% (trace metals basis), foil, Thickness 0.05 mm
Platinum

CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 Solid; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45


  • RTECS: TP2160000
  • Solid

Formula: Pt
Boiling Pt: 3827 °C
Melting Pt: 1772 °C
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR