di-Sodium tetraborate, anhydrous ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Borax, Sodium tetraborate
222825-100G
222825-25G
222825-500G
BT222825-100GEA
224.44
USD
BT222825-100G
BT222825-25G
BT222825-500G
di-Sodium tetraborate, anhydrous ≥99.995%
di-Sodium tetraborate
CAS: 1330-43-4; EC No: 215-540-4; MDL No: MFCD00081185; RTECS: ED4588000
Powder; Linear Formula: Na2B4O7; MW: 201.22
Melting Point: 741°; Boiling Point: 1575° (decomposes)
Density (g/mL): 2.367; Refractive Index: 1.501
Hygroscopic
- RTECS: ED4588000
- Refractive index: 1.501
- Hygroscopic
- Powder
Formula:
Na₂B₄O₇ Boiling Pt: 1575 °C (dec.) Melting Pt: 741 °C Density: 2.367 g/mL |
MDL Number:
MFCD00081185 CAS Number: 1330-43-4 EINECS: 215-540-4 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...