Cobalt ≥99.9%, pieces
Supplier: BeanTown Chemical
222610-100G
222610-2KG
222610-500G
BT222610-100GEA
106.55
USD
BT222610-100G
BT222610-2KG
BT222610-500G
Cobalt ≥99.9%, pieces
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GL5325000
Solid; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92
- RTECS: GL5325000
- Solid
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.92 g/mL |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...