7-Isoquinolinol ≥97%

Supplier: BeanTown Chemical

Synonyms: 7-Hydroxyisoquinoline, Isoquinolin-7-ol

222425-1G 222425-250MG
BT222425-1GEA 161 USD
BT222425-1G BT222425-250MG
7-Isoquinolinol ≥97%
7-Isoquinolinol

CAS: 7651-83-4; MDL No: MFCD00456131 Powder; Molecular Formula: C9H7NO; MW: 145.16 Melting Point: 226-228°


  • Powder

Formula: C₉H₇NO
MW: 145.16
Melting Pt: 226…228 °C
MDL Number: MFCD00456131
CAS Number: 7651-83-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR