Sodium hydrogen carbonate 99,7-100,3 ACS
Supplier: BeanTown Chemical
Synonyms:
Sodium bicarbonate
222160-10KG
222160-2KG
222160-500G
BT222160-10KGEA
163
USD
BT222160-10KG
BT222160-2KG
BT222160-500G
Sodium hydrogen carbonate 99,7-100,3 ACS
Sodium hydrogen carbonate
CAS: 144-55-8; EC No: 205-633-8; MDL No: MFCD00003528; RTECS: VZ0950000
Powder; Linear Formula: NaHCO3; MW: 84.01
Melting Point: 300°
Density (g/mL): 2.16; Refractive Index: 1.500
- RTECS: VZ0950000
- Refractive index: 1.500
- Powder
Formula:
NaHCO₃ MW: 84.01 g/mol Melting Pt: 270 Density: 2.16 g/mL |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 EINECS: 205-633-8 Merck Index: 13,08655 |
|
Specification Test Results
Assay | 99.7-100.3% |
Insoluble matter | ≤0.015% |
Chloride (Cl) | ≤0.003% |
Phosphate (PO4) | ≤0.001% |
Sulfur compounds | ≤0.003% |
Ammonium (NH4) | ≤5ppm |
Heavy metals (as Pb) | ≤5ppm |
Iron (Fe) | ≤0.001% |
Calcium (Ca) | ≤0.02% |
Magnesium (Mg) | ≤0.005% |
Potassium (K) | ≤0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...