β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate, 2-Morpholinoethanesulfonic acid hydrate, 2-(N-Morpholino)ethanesulfonic acid monohydrate
221950-100G
221950-25G
221950-500G
BT221950-100GEA
83.98
USD
BT221950-100G
BT221950-25G
BT221950-500G
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥98%
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
CAS: 145224-94-8; EC No: 224-632-3; MDL No: MFCD00149409; RTECS: QE3479500
Powder; Molecular Formula: C6H13NO4S·H2O; MW: 213.26
Melting Point: 308° (decomposes)
- RTECS: QE3479500
- Powder
Formula:
C₆H₁₃NO₄S·1H₂O Melting Pt: 308 °C (dec.) |
MDL Number:
MFCD00149409 CAS Number: 145224-94-8 EINECS: 224-632-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...