Hexaphenylbenzene ≥98%

Supplier: BeanTown Chemical

221805-1G 221805-5G
BT221805-1GEA 63 USD
BT221805-1G BT221805-5G
Hexaphenylbenzene ≥98%

CAS: 992-04-1; EC No: 213-226-1; MDL No: MFCD00003057 Crystalline; Molecular Formula: C42H30; MW: 534.70 Melting Point: 300°


  • Crystalline

Formula: C₄₂H₃₀
Melting Pt: >300 °C
MDL Number: MFCD00003057
CAS Number: 992-04-1
EINECS: 213-226-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR