Potassium thiocyanate 0.1 N in water

Supplier: BeanTown Chemical

Synonyms: Potassium rhodanide

221760-1L 221760-4L
BT221760-1LEA 47 USD
BT221760-1L BT221760-4L
Potassium thiocyanate 0.1 N in water
Potassium thiocyanate

CAS: 333-20-0; EC No: 206-370-1; MDL No: MFCD00011413 Liquid; Linear Formula: KSCN; MW: 97.18 Melting Point: 173° Density (g/mL): 1.886 Light Sensitive


  • Light Sensitive
  • Liquid

Formula: KSCN
MW: 97.18 g/mol
Boiling Pt: 500
Melting Pt: 175
Density: 1.886 g/mL
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR