Samarium ≥99.9% (trace metals basis excluding Ta), rods Ø 12.7 mm

Supplier: BeanTown Chemical

221620-10MM 221620-25MM 221620-50MM
BT221620-10MMEA 146 USD
BT221620-10MM BT221620-25MM BT221620-50MM
Samarium ≥99.9% (trace metals basis excluding Ta), rods Ø 12.7 mm
Samarium

CAS: 7440-19-9; EC No: 231-128-7; MDL No: MFCD00011233 Solid, cast rod, diameter is approximate; Molecular Formula: Sm; MW: 150.36 Melting Point: 1074°; Boiling Point: 1794° Density (g/mL): 7.47


  • Solid, cast rod, diameter is approximate

Formula: Sm
Boiling Pt: 1794 °C
Melting Pt: 1074 °C
Density: 7.47 g/mL
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR