Tungsten ≥99.95% (trace metals basis), wire Ø 0.025 mm

Supplier: BeanTown Chemical

221360-100M 221360-500M
BT221360-100MEA 91.97 USD
BT221360-100M BT221360-500M
Tungsten ≥99.95% (trace metals basis), wire Ø 0.025 mm
Tungsten

CAS: 7440-33-7; EC No: 231-143-9; MDL No: MFCD00011461 Solid; Molecular Formula: W; MW: 183.84 Melting Point: 3410°; Boiling Point: 5660° Density (g/mL): 19.3


  • Solid

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Density: 19.3 g/mL
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR