Tin ≥99.9999% (trace metals basis), pieces
Supplier: BeanTown Chemical
221260-100G
221260-5G
BT221260-100GEA
698
USD
BT221260-100G
BT221260-5G
Tin ≥99.9999% (trace metals basis), pieces
Tin
CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862; RTECS: XP7320000
Solid; Molecular Formula: Sn; MW: 118.71
Melting Point: 231.9°; Boiling Point: 2270°
Density (g/mL): 7.31
- RTECS: XP7320000
- Solid
Formula:
Sn Boiling Pt: 2270 °C Melting Pt: 231.9 °C Density: 7.31 g/mL |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 |
Specification Test Results
Assay | 99.9999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...