3-(3,4-Dihydroxyphenyl)-DL-alanine ≥96%

Supplier: BeanTown Chemical

Synonyms: DL-β-3,4-Dihydroxyphenyl-Alanine, DL-DOPA, 3,4-Dihydroxy-DL-phenylalanine

221010-100G 221010-25G 221010-5G
BT221010-100GEA 556 USD
BT221010-100G BT221010-25G BT221010-5G
3-(3,4-Dihydroxyphenyl)-DL-alanine ≥96%
3-(3,4-Dihydroxyphenyl)-DL-alanine

CAS: 63-84-3; EC No: 200-566-0; MDL No: MFCD00063060; RTECS: AY5250000 Crystalline; Molecular Formula: C9H11NO4; MW: 197.19 Melting Point: 290° (decomposes)


  • RTECS: AY5250000
  • Crystalline

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 290 °C (dec.)
MDL Number: MFCD00063060
CAS Number: 63-84-3
EINECS: 200-566-0

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR