Dioctyl phenylphosphonate ≥97%

Supplier: BeanTown Chemical

Synonyms: Di-n-octyl phenylphosphonate

220945-25G 220945-5G
BT220945-25GEA 180 USD
BT220945-25G BT220945-5G
Dioctyl phenylphosphonate ≥97%
Dioctyl phenylphosphonate

CAS: 1754-47-8; EC No: 217-142-6; MDL No: MFCD00009561; RTECS: TA0379000 Liquid; Molecular Formula: C22H39O3P; MW: 382.53 Boiling Point: 207°/4 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.967; Refractive Index: 1.478


  • RTECS: TA0379000
  • Refractive index: 1.478
  • Liquid

Formula: C₂₂H₃₉O₃P
MW: 382.53 g/mol
Boiling Pt: 207 °C/4 mmHg
Density: 0.967 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009561
CAS Number: 1754-47-8
EINECS: 217-142-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR