L(+)-Ascorbic acid ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum

220730-100G 220730-25G 220730-500G
BT220730-100GEA 48.77 USD
BT220730-100G BT220730-25G BT220730-500G
L(+)-Ascorbic acid ≥99.0% ACS
L(+)-Ascorbic acid

CAS: 50-81-7; EC No: 200-066-2; MDL No: MFCD00064328; RTECS: CI7650000 Powder; Molecular Formula: C6H8O6; MW: 176.12 Melting Point: 190-194° (decomposes) Optical Rotation: [α]25/D +20.5 to +21.5°, in water


  • RTECS: CI7650000
  • Optical rotation: [α]25/D +20.5 to +21.5°, in water
  • Powder

Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 553
Melting Pt: 190…194 °C (dec.)
MDL Number: MFCD00064328
CAS Number: 50-81-7
EINECS: 200-066-2
Merck Index: 13,00837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR