N-Methyl-2-pyrrolidone (NMP) 99,5 Spectrophotometry

Supplier: BeanTown Chemical

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

220555-1L 220555-4L 220555-4X1L
BT220555-1LEA 81.43 USD
BT220555-1L BT220555-4L BT220555-4X1L
N-Methyl-2-pyrrolidone (NMP) 99,5 Spectrophotometry
N-Methyl-2-pyrrolidone (NMP)

CAS: 872-50-4; EC No: 212-828-1; MDL No: MFCD00003193; RTECS: UY5790000 Liquid; Molecular Formula: C5H9NO; MW: 99.13 Melting Point: -24°; Boiling Point: 202°; Flash point: 91°C (196°F) Density (g/mL): 1.028; Refractive Index: 1.470


  • RTECS: UY5790000
  • Refractive index: 1.470
  • Liquid

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.028 g/mL
Flash Pt: 91 °C (196 °F)
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR