Platinum ≥99.995% (trace metals basis), wire Ø 0.25 mm

Supplier: BeanTown Chemical

Synonyms: Platinium

220450-10CM 220450-250CM 220450-50CM
BT220450-10CMEA 59.81 USD
BT220450-10CM BT220450-250CM BT220450-50CM
Platinum ≥99.995% (trace metals basis), wire Ø 0.25 mm
Platinum

CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 Solid; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45


  • RTECS: TP2160000
  • Solid

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR