(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98
Supplier: BeanTown Chemical
Synonyms:
(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
220380-1G
220380-250MG
BT220380-1GEA
110
USD
BT220380-1G
BT220380-250MG
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 98
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
CAS: 76189-55-4; MDL No: MFCD00010805
Crystalline; Molecular Formula: C44H32P2; MW: 622.67
Melting Point: 239-241°
Optical Rotation: [α]20/D +222°, c = 0.5% in benzene
Air Sensitive
- Optical rotation: [α]20/D +222°, c = 0.5% in benzene
- Air Sensitive
- Crystalline
Formula:
C₄₄H₃₂P₂ MW: 622.69 g/mol Melting Pt: 239…241 °C |
MDL Number:
MFCD00010805 CAS Number: 76189-55-4 Merck Index: 13,01223 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...