Aluminium oxide ≥99% (trace metals basis)

Supplier: BeanTown Chemical

220265-2KG 220265-500G
BT220265-2KGEA 96 USD
BT220265-2KG BT220265-500G
Aluminium oxide ≥99% (trace metals basis)
Aluminium oxide

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 Powder; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • Powder

Formula: Al₂O₃
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR