Artemisinin ≥99%
Supplier: BeanTown Chemical
Synonyms:
Qinghaosu, Arteannuin, (3R,5aS,6R,8aS,9R,12S,12aR)-3,6,9-Trimethyloctahydro-12H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10(3H)-one
220175-1G
220175-5G
BT220175-1GEA
138.79
USD
BT220175-1G
BT220175-5G
Artemisinin ≥99%
Artemisinin
CAS: 63968-64-9; MDL No: MFCD00081057; RTECS: KD4170000
Powder; Molecular Formula: C15H22O5; MW: 282.33
Melting Point: 156-157°
Optical Rotation: [α]20/D +76°, c = 0.5 in methanol
- RTECS: KD4170000
- Optical rotation: [α]20/D +76°, c = 0.5 in methanol
- Powder
Formula:
C₁₅H₂₂O₅ Melting Pt: 156…157 °C |
MDL Number:
MFCD00081057 CAS Number: 63968-64-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...