Magnesium oxide ≥95.0% ACS

Supplier: BeanTown Chemical

Synonyms: Magnesia usta

220135-100G 220135-2KG 220135-500G
BT220135-100GEA 77 USD
BT220135-100G BT220135-2KG BT220135-500G
Magnesium oxide ≥95.0% ACS
Magnesium oxide

CAS: 1309-48-4; EC No: 215-171-9; MDL No: MFCD00011109 Granular; Molecular Formula: MgO ; MW: 40.30 Melting Point: 2852°; Boiling Point: 3600° Density (g/mL): 3.58; Refractive Index: 1.736 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.736
  • Air Sensitive, Moisture Sensitive
  • Granular

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/mL
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Assay 95.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR