Dimethylammonium chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Dimethylamine hydrochloride

220020-1KG 220020-250G 220020-5KG
BT220020-1KGEA 68 USD
BT220020-1KG BT220020-250G BT220020-5KG
Dimethylammonium chloride ≥98%
Dimethylammonium chloride

CAS: 506-59-2; EC No: 208-046-5; MDL No: MFCD00012477; RTECS: IQ0220000 Crystalline; Linear Formula: (CH3)2NH·HCl; Molecular Formula: C2H7N·HCl; MW: 81.54 Melting Point: 170-173° Hygroscopic


  • RTECS: IQ0220000
  • Hygroscopic
  • Crystalline

Formula: C₂H₈ClN
MW: 81.54 g/mol
Melting Pt: 170…173 °C
MDL Number: MFCD00012477
CAS Number: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR