Polyethylene glycol 400

Supplier: BeanTown Chemical

Synonyms: PEG 400

219970-1KG 219970-250G
BT219970-250GEA 41.48 USD
BT219970-250G BT219970-1KG
Polyethylene glycol 400
Polyethylene glycol 400

  • Refractive index: 1.466
  • Liquid

Formula: H(OCH₂CH₂)nOH
MW: 400 g/mol
Melting Pt: 4…8 °C
Density: 1.128 g/mL
Flash Pt: 240 °C (464 °F)
MDL Number: MFCD01779601
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR