Ninhydrin ACS

Supplier: BeanTown Chemical

Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

219340-25G 219340-500G 219340-100G
BT219340-25GEA 70.39 USD
BT219340-25G BT219340-500G BT219340-100G
Ninhydrin ACS
Ninhydrin
Formula: C₉H₆O₄
MW: 178.14 g/mol
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
https://us.vwr.com/stibo/web/std.lang.all/70/01/2297001.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR