Boron carbide ≥99%

Supplier: BeanTown Chemical

219330-500G 219330-100G
BT219330-100GEA 48.49 USD
BT219330-100G BT219330-500G
Boron carbide ≥99%
Boron carbide

  • RTECS: ED7420000
  • <10 Micron Powder

Formula: CB₄
Density: 2.51 g/mL
MDL Number: MFCD00011520
CAS Number: 12069-32-8
EINECS: 235-111-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR