HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

219310-10G 219310-250G 219310-50G
BT219310-10GEA 25.57 USD
BT219310-10G BT219310-250G BT219310-50G
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99%
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)

CAS: 7365-45-9; EC No: 230-907-9; MDL No: MFCD00006158; RTECS: TL6809000 Powder; Molecular Formula: C8H18N2O4S ; MW: 238.31 Refractive Index: 1.339


  • RTECS: TL6809000
  • Refractive index: 1.339
  • Powder

Formula: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR