Sodium bromide ≥99% (dry weight)
Supplier: BeanTown Chemical
219145-5KG
219145-1KG
219145-250G
BT219145-1KGEA
101.09
USD
BT219145-1KG
BT219145-250G
BT219145-5KG
Sodium bromide ≥99% (dry weight)
Sodium bromide
- RTECS: VZ3150000
- Refractive index: 1.641
- Hygroscopic
- Crystalline
Formula:
NaBr MW: 102.89 g/mol Boiling Pt: 1393 °C Melting Pt: 755 °C Density: 3.203 g/mL |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...