Magnesium oxide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Magnesia usta
219095-100G
219095-25G
219095-500G
BT219095-100GEA
189.05
USD
BT219095-100G
BT219095-25G
BT219095-500G
Magnesium oxide ≥99%
Magnesium oxide
CAS: 1309-48-4; EC No: 215-171-9; MDL No: MFCD00011109
40-60nm APS Powder, S.A. 30-40 m2/g; Molecular Formula: MgO ; MW: 40.30
Melting Point: 2852°; Boiling Point: 3600°
Density (g/mL): 3.58; Refractive Index: 1.736
Air Sensitive, Moisture Sensitive
- Refractive index: 1.736
- Air Sensitive, Moisture Sensitive
- 40-60nm APS Powder, S.A. 30-40 m²/g
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 Melting Pt: 2800 Density: 3.58 g/mL |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...