Cesium carbonate ≥99.99%

Supplier: BeanTown Chemical

218930-10G 218930-50G
BT218930-10GEA 58 USD
BT218930-10G BT218930-50G
Cesium carbonate ≥99.99%
Cesium carbonate

CAS: 534-17-8; EC No: 208-591-9; MDL No: MFCD00010957; RTECS: FK9400000 Crystalline; Linear Formula: Cs2CO3; MW: 325.82 Melting Point: 610° (decomposes) Hygroscopic


  • RTECS: FK9400000
  • Hygroscopic
  • Crystalline

Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610 (dec.)
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR