Sodium acetate, anhydrous ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Acetic acid sodium salt
218695-1KG
218695-250G
218695-5KG
BT218695-1KGEA
95.2
USD
BT218695-1KG
BT218695-250G
BT218695-5KG
Sodium acetate, anhydrous ≥99.0% ACS
Sodium acetate
CAS: 127-09-3; EC No: 204-823-8; MDL No: MFCD00012459; RTECS: AJ4300010
Crystalline/Powder; Linear Formula: CH3COONa; MW: 82.03
Melting Point: 300°; Flash point:
- RTECS: AJ4300010
- Hygroscopic
- Crystalline/Powder
Formula:
H₃CCOONa Melting Pt: 300 °C Density: 1.528 g/mL Flash Pt: >250 °C (>482 °F) |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 |
Specification Test Results
Assay | 99.0% min. |
pH of 5% solution at 25°C | 7.5-9.2 |
Loss on drying | ≤1.0% |
Insoluble matter | ≤0.01% |
Phosphate (PO4) | ≤0.001% |
Sulfate (SO4) | ≤0.003% |
Calcium (Ca) | ≤0.005% |
Magnesium (Mg) | ≤0.002% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤0.001% |
Chloride (Cl) | ≤0.002% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...