Ammonium iodide ≥98%

Supplier: BeanTown Chemical

218505-100G 218505-2.5KG 218505-500G
BT218505-100GEA 69.59 USD
BT218505-100G BT218505-2.5KG BT218505-500G
Ammonium iodide ≥98%
Ammonium iodide

CAS: 12027-06-4; EC No: 234-717-7; MDL No: MFCD00011424 Crystalline/Powder; Linear Formula: NH4I ; MW: 144.94 Melting Point: 551° (sublimes) Hygroscopic , Light Sensitive


  • Hygroscopic, Light Sensitive
  • Crystalline/Powder

Formula: NH₄I
MW: 144.94 g/mol
Melting Pt: 551 (subl.)
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 13,00531

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR