Niobium ≥99.95% (trace metals basis excluding Ta), slug, Length 6.35 mm, Diameter 3.175 mm

Supplier: BeanTown Chemical

218485-10G 218485-50G
BT218485-10GEA 38 USD
BT218485-10G BT218485-50G
Niobium ≥99.95% (trace metals basis excluding Ta), slug, Length 6.35 mm, Diameter 3.175 mm
Niobium

CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126 Powder; Molecular Formula: Nb ; MW: 92.91 Melting Point: 2468°; Boiling Point: 4742° Density (g/mL): 8.57


  • Powder

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/mL
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR