Calcium carbonate ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Marble

218130-100G 218130-25G 218130-5G
BT218130-100GEA 529.05 USD
BT218130-100G BT218130-25G BT218130-5G
Calcium carbonate ≥99.995% (trace metals basis)
Calcium carbonate

CAS: 471-34-1; EC No: 207-439-9; MDL No: MFCD00010906; RTECS: FF9335000 Powder; Linear Formula: CaCO3; MW: 100.09 Density (g/mL): 2.93


  • RTECS: FF9335000
  • Powder

Formula: CaCO₃
Density: 2.93 g/mL
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR