Cobalt ≥99.995% (trace metals basis), foil, Thickness 0.25 mm

Supplier: BeanTown Chemical

218080-100X100 218080-50X50MM
BT218080-100X1EA 993 USD
BT218080-100X1 BT218080-50X50
Cobalt ≥99.995% (trace metals basis), foil, Thickness 0.25 mm
Cobalt

CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935 Solid; Molecular Formula: Co; MW: 58.93 Melting Point: 1495°; Boiling Point: 2900° Density (g/mL): 8.92


  • Solid

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.92 g/mL
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR