Albendazole ≥98%

Supplier: BeanTown Chemical

Synonyms: [5-(Propylthio)benzimidazol-2-yl]carbamic Acid Methyl Ester, Methyl [5-(Propylthio)benzimidazol-2-yl]carbamate

217910-100G 217910-10G 217910-50G
BT217910-100GEA 254 USD
BT217910-100G BT217910-10G BT217910-50G
Albendazole ≥98%
Albendazole

CAS: 54965-21-8; EC No: 259-414-7; MDL No: MFCD00083232; RTECS: FD1100000 Solid; Molecular Formula: C12H15N3O2S; MW: 265.33


  • RTECS: FD1100000
  • Solid

Formula: C₁₂H₁₅N₃O₂S
MW: 265.33 g/mol
MDL Number: MFCD00083232
CAS Number: 54965-21-8
EINECS: 259-414-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR