Niobium(V) oxide ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Diniobium pentaoxide, Niobium pentoxide

217865-100G 217865-2KG 217865-500G
BT217865-100GEA 83.23 USD
BT217865-100G BT217865-2KG BT217865-500G
Niobium(V) oxide ≥99.5%
Niobium (V) oxide

CAS: 1313-96-8; EC No: 215-213-6; MDL No: MFCD00011128 -100 Mesh Powder; Molecular Formula: Nb2O5; MW: 265.81 Melting Point: 1520° Density (g/mL): 4.47


  • -100 Mesh Powder

Formula: Nb₂O₅
MW: 265.81 g/mol
Melting Pt: 1496
Density: 4.47 g/mL
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR