Ethyl 4-hydroxybenzoate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
217785-1KG
217785-250G
217785-5KG
BT217785-1KGEA
98
USD
BT217785-1KG
BT217785-250G
BT217785-5KG
Ethyl 4-hydroxybenzoate ≥99%
Ethyl 4-hydroxybenzoate
CAS: 120-47-8; EC No: 204-399-4; MDL No: MFCD00002353; RTECS: DH2190000
Crystalline; Molecular Formula: C9H10O3 ; MW: 166.17
Melting Point: 114-117°; Boiling Point: 297-298°
- RTECS: DH2190000
- Crystalline
Formula:
C₉H₁₀O₃ Boiling Pt: 297…298 °C Melting Pt: 114…117 °C |
MDL Number:
MFCD00002353 CAS Number: 120-47-8 EINECS: 204-399-4 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...